Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106874.1 | NP_001100344.1 | 3 - 4 | 399 | 66 | |
XM_006244305.1 | XP_006244367.1 | 3 - 4 | 448 | 66 | ||
XM_008766754.1 | XP_008764976.1 | 3 - 4 | 509 | 66 | ||
GenBank mRNA | FM165287.1 | - | 3 - 4 | 336 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |