Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107835.2 | NP_001101305.1 | 5 - 6 | 1049 | 127 | |
XM_006233976.1 | XP_006234038.1 | 5 - 6 | 1183 | 127 | ||
XM_006233977.2 | XP_006234039.1 | 6 - 7 | 1099 | 127 | ||
XM_006233980.2 | XP_006234042.1 | 6 - 7 | 1007 | 127 | ||
XM_008761750.1 | XP_008759972.1 | 6 - 7 | 2658 | 127 | ||
XM_008761751.1 | XP_008759973.1 | 7 - 8 | 1264 | 127 | ||
XM_008761752.1 | XP_008759974.1 | 5 - 6 | 985 | 127 | ||
GenBank mRNA | BC166990.1 | - | 5 - 6 | 1049 | 7110795 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |