Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Surf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033868.1 | NP_001029040.1 | 1 - 2 | 166 | 64 | |
GenBank mRNA | CB703892.1 | - | 1 - 2 | 178 | 64 | |
CK358924.1 | - | 1 - 2 | 100 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |