Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012094.1 | NP_001012094.1 | 1 - 2 | 97 | 93 | |
XM_006233973.2 | XP_006234035.1 | 1 - 2 | 166 | 93 | ||
GenBank mRNA | BC087042.1 | - | 1 - 2 | 97 | 7107163 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |