Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106562.1 | NP_001100032.1 | 9 - 10 | 1447 | 79 | |
XM_006233757.2 | XP_006233819.1 | 9 - 10 | 1438 | 79 | ||
XM_008761634.1 | XP_008759856.1 | 9 - 10 | 1354 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Transporter 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |