Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107942.2 | NP_001101412.2 | 6 - 7 | 1288 | 117 | |
GenBank mRNA | BC167044.1 | - | 4 - 5 | 686 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |