Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006249749.2 | XP_006249811.1 | 1 - 2 | 282 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |