Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751244.2 | XP_003751292.1 | 17 - 18 | 1382 | 98 | |
XM_003751245.2 | XP_003751293.1 | 18 - 19 | 1442 | 98 | ||
XM_006249933.2 | XP_006249995.1 | 17 - 18 | 1452 | 98 | ||
XM_006249934.1 | XP_006249996.1 | 18 - 19 | 1369 | 98 | ||
XM_008769580.1 | XP_008767802.1 | 16 - 17 | 1272 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |