Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106098.2 | NP_001099568.2 | 8 - 9 | 696 | 112 | |
XM_006253493.2 | XP_006253555.1 | 8 - 9 | 1680 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |