Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107576.1 | NP_001101046.1 | 1 - 2 | 151 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus 1700 Probe spans exons |