Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191988.1 | NP_001178917.1 | 2 - 3 | 250 | 90 | |
GenBank mRNA | DQ630444.1 | - | 2 - 3 | 229 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |