Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108101.1 | NP_001101571.1 | 3 - 4 | 554 | 110 | |
GenBank mRNA | BC166731.1 | - | 3 - 4 | 629 | 7934689 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |