Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135582.1 | NP_001129054.1 | 4 - 5 | 306 | 80 | |
GenBank mRNA | FQ219839.1 | - | 3 - 4 | 275 | 80 | |
FQ220026.1 | - | 3 - 4 | 275 | 80 | ||
FQ224899.1 | - | 3 - 4 | 275 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |