Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amelx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271073.1 | NP_001258002.1 | 4 - 5 | 215 | 62 | |
GenBank mRNA | U51195.1 | - | 4 - 5 | 134 | ||
U60562.1 | - | 4 - 5 | 216 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |