Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amelx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271073.1 | NP_001258002.1 | 6 - 7 | 623 | 107 | |
NM_001271074.1 | NP_001258003.1 | 6 - 7 | 695 | 107 | ||
NM_001271075.1 | NP_001258004.1 | 6 - 7 | 332 | 107 | ||
GenBank mRNA | U51195.1 | - | 6 - 7 | 696 | 107 | |
U60562.1 | - | 6 - 7 | 624 | 107 | ||
U60564.1 | - | 6 - 7 | 328 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |