Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zic3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109019.2 | NP_001102489.1 | 1 - 2 | 972 | 79 | |
XM_006257586.2 | XP_006257648.1 | 2 - 3 | 1365 | 79 | ||
XM_008773570.1 | XP_008771792.1 | 2 - 3 | 1385 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |