Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109503.1 | NP_001102973.1 | 1 - 2 | 131 | 56 | |
GenBank mRNA | BC098743.1 | - | 1 - 2 | 106 | 56 | |
FQ229674.1 | - | 1 - 2 | 75 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |