Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ccrn4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138526.1 | NP_612535.1 | 1 - 2 | 408 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |