Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271383.1 | NP_001258312.1 | 6 - 7 | 726 | 63 | |
XM_008763851.1 | XP_008762073.1 | 6 - 7 | 746 | 63 | ||
XM_008763852.1 | XP_008762074.1 | 6 - 7 | 687 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |