Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750410.3 | XP_003750458.1 | 2 - 3 | 755 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |