Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csrnp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108113.1 | NP_001101583.1 | 2 - 3 | 692 | 64 | |
XM_006242337.2 | XP_006242399.1 | 1 - 2 | 235 | 64 | ||
GenBank mRNA | FQ223052.1 | - | 2 - 3 | 456 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Recommended Assay Rattus norvegicus Probe spans exons |