Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172063.2 | NP_001165534.1 | 7 - 8 | 1504 | 68 | |
XM_006242294.2 | XP_006242356.1 | 9 - 10 | 2590 | 68 | ||
XM_006242295.2 | XP_006242357.1 | 9 - 10 | 1913 | 68 | ||
XM_006242296.2 | XP_006242358.1 | 9 - 10 | 1857 | 68 | ||
XM_008765651.1 | XP_008763873.1 | 10 - 11 | 2092 | 68 | ||
XM_008765652.1 | XP_008763874.1 | 9 - 10 | 1849 | 68 | ||
XM_008765653.1 | XP_008763875.1 | 9 - 10 | 2129 | 68 | ||
GenBank mRNA | FQ211776.1 | - | 9 - 10 | 1882 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |