Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760810.1 | XP_008759032.1 | 3 - 4 | 348 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |