Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008722.1 | NP_001008722.1 | 6 - 7 | 979 | 60 | |
XM_006255737.2 | XP_006255799.1 | 7 - 8 | 1027 | 60 | ||
GenBank mRNA | AB120371.1 | - | 6 - 7 | 979 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |