Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025765.1 | NP_001020936.1 | 3 - 4 | 298 | 66 | |
XM_006255565.2 | XP_006255627.1 | 1 - 2 | 1781 | 66 | ||
XM_006255566.2 | XP_006255628.1 | 2 - 3 | 363 | 66 | ||
XM_006255567.2 | XP_006255629.1 | 3 - 4 | 449 | 66 | ||
GenBank mRNA | BC097467.1 | - | 3 - 4 | 298 | 7457672 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |