Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010947.1 | NP_001010947.1 | 4 - 5 | 991 | 70 | |
GenBank mRNA | BC089060.1 | - | 4 - 5 | 991 | 7320396 | 70 |
FQ213644.1 | - | 4 - 5 | 983 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |