Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107707.1 | NP_001101177.1 | 11 - 12 | 1628 | 82 | |
XM_006233050.2 | XP_006233112.1 | 10 - 11 | 4022 | 82 | ||
GenBank mRNA | FQ211559.1 | - | 7 - 8 | 1209 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Rattus norvegicus Probe spans exons |