Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mab21l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106456.1 | NP_001099926.1 | 4 - 5 | 925 | 76 | |
XM_006233045.2 | XP_006233107.1 | 4 - 5 | 836 | 76 | ||
XM_006233046.2 | XP_006233108.1 | 4 - 5 | 1120 | 76 | ||
XM_008761374.1 | XP_008759596.1 | 4 - 5 | 817 | 76 | ||
XM_008761375.1 | XP_008759597.1 | 4 - 5 | 741 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |