Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gimap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011968.3 | NP_001011968.1 | 2 - 3 | 432 | 64 | |
GenBank mRNA | AJ633683.1 | - | 2 - 3 | 435 | 64 | |
BC089859.1 | - | 341 | 7384582 | 64 | ||
DQ125342.1 | - | 2 - 3 | 342 | 64 | ||
DQ125343.1 | - | 342 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |