Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108318.2 | NP_001101788.1 | 13 - 14 | 1693 | 97 | |
XM_006248348.2 | XP_006248410.1 | 13 - 14 | 1894 | 97 | ||
XM_006248349.2 | XP_006248411.1 | 13 - 14 | 1885 | 97 | ||
GenBank mRNA | FQ211993.1 | - | 13 - 14 | 1726 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Ion Channel Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |