Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213630.1 | NP_998795.1 | 4 - 5 | 623 | 74 | |
XM_006248341.2 | XP_006248403.1 | 4 - 5 | 855 | 74 | ||
GenBank mRNA | AY590499.1 | - | 4 - 5 | 623 | 74 | |
BC092588.1 | - | 4 - 5 | 791 | 7368263 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |