Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107093.1 | NP_001100563.1 | 2 - 3 | 420 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |