Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hlcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248111.2 | XP_006248173.1 | 9 - 10 | 3095 | 127 | |
XM_006248112.2 | XP_006248174.1 | 9 - 10 | 3162 | 127 | ||
XM_006248114.2 | XP_006248176.1 | 8 - 9 | 2956 | 127 | ||
XM_008768611.1 | XP_008766833.1 | 9 - 10 | 2294 | 127 | ||
XM_008768612.1 | XP_008766834.1 | 8 - 9 | 2159 | 127 | ||
XM_221630.8 | XP_221630.6 | 8 - 9 | 2275 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |