Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191759.1 | NP_001178688.1 | 2 - 3 | 251 | 68 | |
XM_006238495.2 | XP_006238557.1 | 3 - 4 | 571 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |