Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006969.1 | NP_001006970.1 | 3 - 4 | 476 | 82 | |
XM_006229021.1 | XP_006229083.1 | 3 - 4 | 1078 | 82 | ||
XM_006229022.2 | XP_006229084.1 | 3 - 4 | 1038 | 82 | ||
XM_006229023.2 | XP_006229085.1 | 3 - 4 | 597 | 82 | ||
XM_006229024.2 | XP_006229086.1 | 3 - 4 | 465 | 82 | ||
GenBank mRNA | BC083776.1 | - | 3 - 4 | 476 | 7193483 | 82 |
FQ213393.1 | - | 3 - 4 | 612 | 82 | ||
FQ218848.1 | - | 3 - 4 | 624 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |