Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001076463.5 | XP_001076463.2 | 3 - 4 | 425 | 66 | |
XM_006229208.2 | XP_006229270.1 | 2 - 3 | 894 | 66 | ||
XM_008759458.1 | XP_008757680.1 | 3 - 4 | 353 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |