Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013088.1 | NP_001013106.1 | 5 - 6 | 695 | 89 | |
XM_008763594.1 | XP_008761816.1 | 5 - 6 | 1060 | 89 | ||
XM_008763595.1 | XP_008761817.1 | 4 - 5 | 998 | 89 | ||
GenBank mRNA | BC079327.1 | - | 5 - 6 | 695 | 7135490 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |