Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008513.1 | NP_001008513.1 | 3 - 4 | 408 | 74 | |
GenBank mRNA | BC086571.1 | - | 3 - 4 | 408 | 7304213 | 74 |
FQ218040.1 | - | 3 - 4 | 432 | 74 | ||
FQ223815.1 | - | 3 - 4 | 434 | 74 | ||
FQ225478.1 | - | 3 - 4 | 432 | 74 | ||
FQ226079.1 | - | 3 - 4 | 430 | 74 | ||
FQ233622.1 | - | 3 - 4 | 432 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Rat Chemokine Panel 1700 Inventoried Probe spans exons |