Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039009.1 | NP_001034098.1 | 2 - 3 | 349 | 105 | |
XM_008763765.1 | XP_008761987.1 | 2 - 3 | 3251 | 105 | ||
XM_008763766.1 | XP_008761988.1 | 3 - 4 | 547 | 105 | ||
XM_008763767.1 | XP_008761989.1 | 3 - 4 | 406 | 105 | ||
XM_008763768.1 | XP_008761990.1 | 3 - 4 | 2960 | 105 | ||
GenBank mRNA | AY886536.1 | - | 2 - 3 | 349 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |