Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013227.2 | NP_001013245.1 | 3 - 4 | 253 | 71 | |
GenBank mRNA | BC063182.1 | - | 3 - 4 | 257 | 6887869 | 71 |
FQ215552.1 | - | 3 - 4 | 252 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |