Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053577.1 | NP_446029.1 | 218 | 66 | ||
GenBank mRNA | BC088193.1 | - | 218 | 7372900 | 66 | |
FQ210158.1 | - | 134 | 66 | |||
FQ210198.1 | - | 155 | 66 | |||
FQ210350.1 | - | 182 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |