Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133412.1 | NP_596903.1 | 1 - 2 | 547 | 147 | |
XM_006245576.2 | XP_006245638.1 | 1 - 2 | 617 | 147 | ||
GenBank mRNA | AF321415.1 | - | 1 - 2 | 547 | 147 |
Gene Symbol: LOC102554121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245586.2 | XP_006245648.1 | 1 - 2 | 618 | 147 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Rattus norvegicus Probe spans exons |