Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trdmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031643.1 | NP_001026813.1 | 5 - 6 | 426 | 155 | |
GenBank mRNA | BC098700.1 | - | 5 - 6 | 426 | 7387075 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |