Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnpy4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108852.2 | NP_001102322.1 | 4 - 5 | 491 | 68 | |
GenBank mRNA | BC162041.1 | - | 4 - 5 | 491 | 7100495 | 68 |
FQ226241.1 | - | 4 - 5 | 510 | 68 | ||
FQ229096.1 | - | 4 - 5 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |