Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108633.1 | NP_001102103.1 | 5 - 6 | 556 | 64 | |
XM_008762991.1 | XP_008761213.1 | 6 - 7 | 525 | 64 | ||
XM_008762992.1 | XP_008761214.1 | 5 - 6 | 597 | 64 | ||
XM_008762993.1 | XP_008761215.1 | 6 - 7 | 525 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Rattus norvegicus Probe spans exons |