Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slain2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107214.1 | NP_001100684.1 | 7 - 8 | 1685 | 84 | |
XM_006250898.2 | XP_006250960.1 | 7 - 8 | 1760 | 84 | ||
GenBank mRNA | BC158815.1 | - | 5 - 6 | 758 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |