Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004217.4 | NP_001004217.2 | 4 - 5 | 698 | 94 | |
GenBank mRNA | AY621629.1 | - | 3 - 4 | 502 | 94 | |
BC097929.1 | - | 4 - 5 | 698 | 7444887 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |