Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004217.4 | NP_001004217.2 | 5 - 6 | 824 | 122 | |
GenBank mRNA | AY621629.1 | - | 4 - 5 | 628 | 122 | |
BC097929.1 | - | 5 - 6 | 824 | 7444887 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |