Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023955.1 | NP_076445.1 | 4 - 5 | 435 | 60 | |
GenBank mRNA | AF295405.1 | - | 3 - 4 | 279 | 60 | |
BC085862.1 | - | 4 - 5 | 435 | 7190371 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |