Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023955.1 | NP_076445.1 | 7 - 8 | 833 | 73 | |
GenBank mRNA | AF295405.1 | - | 6 - 7 | 677 | 73 | |
BC085862.1 | - | 7 - 8 | 833 | 7190371 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |